Would you like to log in again?
VWR® Round-Bottom Boiling Flasks
Supplier: VWR
057.202.58A 057.202.59 057.202.61 057.202.59A 057.202.60 057.202.58 057.202.57 057.202.62
10536-654CS 257.87 USD InStock
10536-654 10536-664 10536-666 10536-668 10536-670 10536-672 10536-652 10536-674
VWR® Round-Bottom Boiling Flasks
Flasks Round Bottom Flasks
Durable boiling flasks have good temperature and chemical resistance.
  • Good temperature resistance
  • Chemically resistant
  • Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403 Type 1. The flasks feature uniform wall thickness, which makes them ideal for heating.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty