Would you like to log in again?
VWR® Volumetric Pipettes, Reusable, Color Coded, Class A
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
123.450.03 123.450.03A 123.450.06B 123.450.03D 123.450.04A 123.450.05 123.450.00 123.450.03C 123.450.08 123.450.01 123.450.02A 123.450.02 123.450.06 123.450.02B 123.450.03B 123.450.04 123.450.06A 123.450.07
10546-018CS 175.99 USD InStock
10546-018 10546-008 10546-016 10546-038 10546-028 10546-006 10546-014 10546-036 10546-026 10546-004 10546-034 10546-012 10546-024 10546-010 10546-032 10546-022 10546-030 10546-020
VWR® Volumetric Pipettes, Reusable, Color Coded, Class A
Pipettes Serological Pipettes
Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E969.
  • Reusable
  • Color-coded for easy sorting
  • Borosilicate glass fabrication
Calibrated at 20 °C TD-EX. These pipettes are unserialized or batch certified.
Certifications: Pipettes are color coded as per ASTM specification E1273.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty