Would you like to log in again?
VWR® Powder Funnels, Short Stem
Supplier: VWR
238.404.10P 238.404.08P 238.404.12P 238.404.15P 238.404.06P
10546-050CS 408.86 USD InStock
10546-050 10546-048 10545-910 10546-046 10545-912
VWR® Powder Funnels, Short Stem
Funnels Powder Funnels
Funnels comes with short, wide stem, and are useful for adding powder chemical to reaction vessels.
  • Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass
  • Constructed with a heavy, uniform wall
  • Fire-polished rim and stem to provide long service life
Designed from ASTM Specification E1095 requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty