Would you like to log in again?
SP Bel-Art Single Tube Holder Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
F18794-0003
10600-006EA 29.42 USD InStock
10600-006
SP Bel-Art Single Tube Holder Racks, Bel-Art Products, a part of SP
Racks Tube Racks
This holder solves the problem of where to put a single 50 mL conical tube where it will not roll or spill.
  • Epoxy-coated stainless steel construction
  • Chemical- and corrosion-resistant
  • Autoclavable
Steel wire with epoxy coating is chemical and corrosion resistant and steam autoclavable at 121°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty