Would you like to log in again?
SURGIFOAM® Absorbable Gelatin Oral Sponge, ETHICON
Supplier: Ethicon (Johnson & Johnson)
ETH1974 ETH1972 ETH1977
10611-590CS 717.65 USD InStock
10611-590 10611-586 10611-594
SURGIFOAM® Absorbable Gelatin Oral Sponge, ETHICON
Wipes Sponges
The SURGIFOAM® absorbable gelatin sponge, U.S.P. is a sterile, water-insoluble, malleable, porcine gelatin absorbable sponge intended for hemostatic use by applying to a bleeding surface. The sponge is off-white and porous in appearance.
  • Absorbs completely within 4 to 6 weeks, with little tissue reaction
The oral sponge can be used in oral surgery and slightly compressed to secure hemostatic effect in cavities after tooth extractions etc. May also be used in areas where the small size of the sponge is an advantage, e.g., Epistaxis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty