Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alkaline Phosphatase Assay Kit

Supplier: Abnova
KA1642
10626-232EA 653.85 USD InStock
10626-232
Alkaline Phosphatase Assay Kit
Assays Enzyme Assays
Alkaline phosphatase assay kit allows the colorimetric kinetic determination of serum alkaline phosphatase activity.

  • High sensitivity and wide linear range: Use 5 µl serum or plasma sample; detection limit is 2 U/L, linear up to 800 U/L
  • Homogeneous and simple procedure: Simple 'mix-and-measure' procedure allows reliable quantitation of ALP activity within 5 minutes
  • Robust and amenable to HTS: All reagents are compatible with high-throughput liquid handling instruments

Applications:
Direct Assays: ALP activity in serum, plasma and other sources
Characterisation and quality control for ALP production
Drug Discovery: High throughput screening for ALP inhibitors and evaluation of ALP inhibitors

Delivery information: Kit contains assay buffer 50 ml (pH 10.5), Mg Acetate 1.5 ml (0.2 M), pNPP liquid 600 µl (1 M) and Tartrazine calibrator 10 ml.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty