Would you like to log in again?
Antioxidant Assay Kit
Supplier: Abnova
KA1622
10628-438EA 578.46 USD InStock
10628-438
Antioxidant Assay Kit
Assays Cellular Assays
Antioxidant assay kit is a quantitative colorimetric determination of total antioxidant capacity of cellular samples.
  • Sensitive and accurate, linear detection range from 1.5 to 1000 µM Trolox equivalents
  • Simple procedure involves addition of a single working reagent and incubation for 10 min
  • Can be readily automated as a high-throughput assay for thousands of samples per day
Antioxidant assay kit measures total antioxidant capacity in which Cu²⁺ is reduced by antioxidant to Cu⁺. The resulting Cu⁺ specifically forms a colored complex with a dye reagent. The color intensity at 570 nm is proportional to TAC in the sample.
Antioxidant assay kit is suitable for serum, plasma, urine, saliva and other biological samples, food and beverages. Also used in drug discovery/pharmacology to check the effect of drugs on TAC.
Delivery information: Kit includes Reagent A, 12 ml; Reagent B, 1 ml; and Standard (50 mM Trolox, 100 µl).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty