Would you like to log in again?
Cotinine ELISA Kit, HRP-Conjugate, Abnova
Supplier: Abnova
KA0930
10630-348EA 507.69 USD InStock
10630-348
Cotinine ELISA Kit, HRP-Conjugate, Abnova
Assays ELISAs
Cotinine ELISA kit is used for the measurement of Cotinine in serum and urine.
  • Solid phase, competitive ELISA
  • Cotinine is a reliable marker for smoking cessation studies
  • Detection of Cotinine in serum and urine
Exposure to tobacco smoke can be detected by measuring nicotine and its metabolites
The Cotinine ELISA Kit is a solid phase competitive ELISA. The samples and Cotinine enzyme conjugate are added to the wells coated with anti-Cotinine antibody. Cotinine in the samples competes with a Cotinine enzyme (HRP) conjugate for binding sites. Unbound Cotinine and Cotinine enzyme conjugate is washed off by washing step. Upon the addition of the substrate, the intensity of color is inversely proportional to the concentration of Cotinine in the samples. A standard curve is prepared relating color intensity to the concentration of the Cotinine.
Nicotine has a short half-life and is not used as a marker for tobacco smoke exposure. Cotinine due to its longer half-life has been used in research as a reliable marker for smoking status and smoking cessation studies. The Cotinine ELISA Kit is designed for the detection Cotinine in serum and urine. It can also be adapted for other fluids.
Delivery information: Kit contains microwell coated with polyclonal Ab to Cotinine (12x8x1); ready to use Standard set (0.5 mL x6
); Cotinine HRP enzyme conjugate (12 mL
); TMB substrate (12 mL
) and Stop solution (12 mL
).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty