Would you like to log in again?
Corticosterone ELISA Kit, Abnova
Supplier: Abnova
KA0468
10630-576EA 653.85 USD InStock
10630-576
Corticosterone ELISA Kit, Abnova
Assays ELISAs
Corticosterone ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of corticosterone.
  • Suitable for cell culture samples, milk, plasma, saliva, serum, urine
Limit of detection: 0.3 ng/ml.
Caution: For research use only (RUO).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty