Would you like to log in again?
Dipase (from Bacillus polymyxa)
Supplier: Abnova
P5318
10630-956EA 316.92 USD InStock
10630-956
Dipase (from Bacillus polymyxa)
Enzymes
Dispase is a mild protease which cleaves fibronectin, collagen IV and to a lesser extent collagen I.
Lyophilized and partially purified dispase is used for Neutral Protease (Dispase) Assay, tissue disaggregation and subcultivation, prevention of unwanted cell clumping, preparation of cells for culture, separation of intact epidermis from dermis and intact epithelial sheet in culture from the substratum, harvest and transfer of cell lines, gentle and intact detachment of epidermal cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty