Would you like to log in again?
Insulin (Mouse) ELISA Kit, Abnova
Supplier: Abnova
KA3812
10633-576EA 507.69 USD InStock
10633-576
Insulin (Mouse) ELISA Kit, Abnova
Assays ELISAs
Insulin (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse insulin.
  • Calibration Range: 0 to 140 uIU/mL
  • Label: HRP-conjugated
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty