Would you like to log in again?
LDL Uptake Assay Kit (Cell-Based), Abnova
Supplier: Abnova
KA1327
10633-926EA 653.85 USD InStock
10633-926
LDL Uptake Assay Kit (Cell-Based), Abnova
Assays Cellular Assays
LDL uptake cell-based assay kit is a tool for studying LDL uptake and regulation at the cellular level.
Delivery information: Kit includes cell-based assay fixative, rabbit anti-LDL receptor primary antibody, cell-based assay blocking solution, LDL-DyLightTM 550 and DyLightTM 488-conjugated goat anti- rabbit IgG secondary antibody.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty