Would you like to log in again?
Thiol Assay Kit, Abnova
Supplier: Abnova
KA4133
10636-374EA 363.08 USD InStock
10636-374
Thiol Assay Kit, Abnova
Assays Cellular Assays
Thiol assay kit (green fluorescence) is a fluorometric method for the quantification of thiol groups in a variety of molecules such as small molecules or proteins.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty