Would you like to log in again?
Acriflavin-Cefsulodin-Vancomycin (ACV) Supplement, Himedia®
Supplier: HiMedia Laboratories
FD284-5VL FD284-5VL
10662-034PK 274.62 USD InStock
CA10662-034 10662-034
Acriflavin-Cefsulodin-Vancomycin (ACV) Supplement, Himedia®
Media for Microbiology Additives and Supplements for Microbiology
ACV supplement is used for the isolation of Enterohemorrhagic <italic>E
One vial is sufficient for 500 mL medium.
coli</italic> (EHEC).
Caution: Store at 2 to 8 °C. For medicinal use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty