Would you like to log in again?
TitraLab® AT1000 Potentiometric Titrator, 2 Burettes, 2 Pumps
Supplier: Hach
TitraLab®
AP0011.AT1222 AP0013.AT1222 AP0011.AT1222 AT122297 AP0005.AT1222 AP0005.AT1222 AT1222.97 AP0013.AT1222 AT1222.97
10662-204EA 3168.75 USD InStock
CA10662-204 10662-204 CA10662-258 CA10662-200 10662-200 10662-258 104085-116 CA10662-172 10662-172
TitraLab® AT1000 Potentiometric Titrator, 2 Burettes, 2 Pumps
Titrators
Automatic titration without all the complications.
  • Reliable titration results
  • Eliminates operator interpretation and manual processes
  • Simple setup and titration
  • Application-specific functions
Simplify complex titration with the TitraLab® AT1000 from Hach.
Eliminates operator interpretation and manual processes with automatic titration to quickly deliver accurate and repeatable results. Pre-programmed titration methods detect end points and eliminate manual calculations to make results easier to achieve without advanced programming. Application-specific functions eliminate complex titration setup and analysis. Hach’s unique application kits make it quick for anyone to set up and operate a test.
Recommended Application Packages include Water: pH, alkalinity and hardness (ISE).
Food add beverage: pH, total acidity and SO₂ in wine.
Food and beverage: pH, total acidity and salt.
Application packages for use with TitraLab 1000 Series titrators from Hach contain all elements but reagents to make it quick and easy for anyone to set up and operate a test. Each application Package is specifically designed for unique parameters to ensure accurate and reproducible measurements at glance. Packages include adapters, glass bottle, magnetic stirring bars, polypropylene beakers, bottle stopper with desiccant tube, electrode(s) (Digital IntelliCAL® or analogue), high precision glass syringe (Hamilton) and USB key containing application notes and applications to automatically program titrators for selected parameters. Setting up your application has never been so simple.
Certifications: Safety IEC/EN 61010-1; EMC IEC/EN 61326-1. Manufacturing environment: FDA cGMP, ISO 9001 and ISO 13485 Compliant Quality System.
Accessories information: No applications kits or accessories included.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty