Would you like to log in again?
Cyto-ID® Autophagy Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51031-0050 ENZ-51031-0050 ENZ-51031-K200
10662-266EA 390.92 USD InStock
CA10662-266 10662-266 89165-926
Cyto-ID® Autophagy Detection Kit, Enzo Life Sciences
Assays Cellular Assays
The Cyto-ID® Autophagy detection kit provides a rapid, specific and quantitative approach for monitoring autophagy in live cells by fluorescence microscopy and flow cytometry.
Cyto-ID® Autophagy Detection Kit measures autophagic vacuoles and monitors autophagic flux in live cells using a novel dye that selectively labels autophagic vacuoles. The dye has been optimized through the identification of titratable functional moieties that allow for minimal staining of lysosomes while exhibiting bright fluorescence upon incorporation into pre-autophagosomes, autophagosomes, and autolysosomes (autophagolysosomes).
Delivery information: Kit contains Cyto-ID® Green Detection Reagent, 50 µL, Hoechst 33342 Nuclear Stain, 50 µL Autophagy Inducer (Rapamycin, 25 nmol) and 10X Assay Buffer, 30 mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty