Would you like to log in again?
Amersham ECL Western Blotting Start Detection Reagents, Cytiva
Supplier: Cytiva
29489438 RPN3244 RPN3243 RPN3244 RPN3243
10662-310EA 215.93 USD InStock
CA10746-452 MPCA29489438 10662-310 10662-312 CA10746-470
Amersham ECL Western Blotting Start Detection Reagents, Cytiva
Electrophoresis Reagents Western Blotting Reagents
Chemiluminescence detection reagent for detection of high to medium abundant proteins.
  • For outine and confirmatory western blotting, high expressed proteins and tagged proteins
  • Detection with X-ray film or CCD Imager
  • Stable until the expiration date printed on box when stored under recommended conditions
  • Signal duration < 3 h
Recommended detection with Amersham Hyperfilm™ (X-ray film) or ImageQuant™ LAS 500. Optimal membrane amersham protran.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty