Would you like to log in again?
PROTEOSTAT® Aggresome Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51035-0025 ENZ-51035-K100
10662-314EA 155 USD InStock
10662-314 89409-190
PROTEOSTAT® Aggresome Detection Kit, Enzo Life Sciences
Assays Protein Assays
Robust, quantitative detection of aggresomes relevant to the study of neurodegenerative disease, liver disease and toxicology.
The ProteoStat®Aggresome detection kit assay is potentially applicable to the identification of small molecules that inhibit aggresome formation as well as immuno-localization studies between aggregated protein cargo and the various proteins implicated in aggresome formation, such as histone deacetylase 6, parkin, ataxin-3, dynein motor complex and ubiquilin-1.
Aggresomes are inclusion bodies that form when the ubiquitin–proteasome machinery is overwhelmed with aggregation-prone proteins. Typically, an aggresome forms in response to some cellular stress, such as hyperthermia, viral infection, or exposure to reactive oxygen species. Aggresomes may provide a cytoprotective function by sequestering the toxic, aggregated proteins and may also facilitate their ultimate elimination from cells by autophagy.Certain cellular inclusion bodies associated with human disease are thought to arise from an aggresomal response including Alzheimer’s disease as it relates to intracellular and extracellular aggregates, Lewy bodies associated with neurons in Parkinson's disease, Mallory bodies associated with liver cells in alcoholic liver disease and Hyaline inclusion bodies associated with astrocytes in amyotrophic lateral sclerosis (ALS).ProteoStat® Aggresome Detection kit provides a rapid, specific and quantitative approach to identify inhibitors relevant to neurodegenerative disease in an authentic cellular context. This fixed-cell assay doesn’t require non-physiological protein mutations or genetically engineered cell lines. ProteoStat® dye has been validated under a wide range of conditions and with small molecule modulators, demonstrating suitability for screening compounds of potential therapeutic value, and optimized for antibody co-localization studies to identify interactions between aggregated protein cargo and the various proteins implicated in autophagy and aggresome formation.
Ordering information: Includes ProteoStat®Aggresome detection reagent, 10µl, Hoechst 33342 Nuclear stain, 50µl, Proteasome Inhibitor (MG-132), 120 nmol, 10X Assay Buffer, 25ml.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty