Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDZD7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-9042R-A488
10666-040EA 792.04 USD InStock
10666-040
Anti-PDZD7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
PDZK7, also known as PDZD7, is a 517 amino acid protein that contains two PDZ (DHR) domains. Encoded by a gene that maps to human chromosome 10q24.31, PDZK7 is conserved in dog, mouse and rat, and exists as three alternatively spliced isoforms. PDZK7 is known to interact with Harmonin, MASS1, USH1G and Usherin. Localizing to nucleus, PDZK7 is expressed in retinal pigment epithelium and inner ear. Biallelic inactivation of PDZK7 can cause non-syndromic hearing impairment and chromosomal aberrations, which are linked to non-syndromic sensorineural deafness. PDZK7 mutations are also linked to Usher syndrome, which is characterized by retinitis pigmentosa and sensorineural deafness, and Alzheimer disease. The gene that encodes PDZK7 maps to human chromosome 10q24.31.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDZD7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty