Would you like to log in again?
VWR® C-Leg Bench Frame with Top for Single Bay Uprights
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
14-C101VWR487 14-C103VWR490 14-C100VWR491 14-C101VWR490 14-C100VWR482 14-C103VWR488 14-C100VWR485 14-C103VWR487 14-C103VWR482 14-C101VWR486 14-C103VWR485 14-C100VWR489 14-C100VWR481 14-C103VWR483 14-C103VWR489 14-C101VWR491 14-C103VWR480 14-C103VWR481 14-C100VWR490 14-C101VWR481 14-C101VWR480 14-C100VWR487 14-C103VWR484 14-C100VWR480 14-C101VWR485 14-C101VWR482 14-C101VWR484 14-C103VWR491 14-C100VWR488 14-C100VWR483 14-C100VWR486 14-C101VWR483 14-C101VWR489 14-C100VWR484 14-C103VWR486 14-C101VWR488
10682-618EA 2044.14 USD InStock
10682-618 10682-662 10682-640 10682-584 10682-660 10682-582 10682-580 10682-626 10682-646 10682-624 10682-568 10682-622 10682-644 10682-666 10682-664 10682-620 10682-642 10682-628 10682-652 10682-630 10682-772 10682-574 10682-650 10682-572 10682-770 10682-570 10682-638 10682-636 10682-658 10682-656 10682-634 10682-578 10682-654 10682-632 10682-774 10682-576
VWR® C-Leg Bench Frame with Top for Single Bay Uprights
Furniture Tables and Workbenches
These VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads (static) up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. All bench frames can be equipped with casters to create a mobile workstation solution. Frame color is light neutral.
Ordering information: VWR® lab benches are height adjustable in manual (without hand crank), hand crank, and motorized versions. Configurations include frame (pair of legs, rear case, and front rail) and top for single bay uprights (uprights not included). Top includes black phenolic or black laminate options.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty