Would you like to log in again?
VWR® C-Leg C-Leg Bench Frames
Supplier: VWR
100VWR063P 105VWR063P 100VWR033P 101VWR065 101VWR063 105VWR065P 105VWR033P 101VWR032P 100VWR031P 101VWR033P 105VWR064P 101VWR064 105VWR032P 105VWR031P 100VWR032P 100VWR064P 100VWR065P 101VWR031P
10682-762EA 4334.4 USD InStock
10682-762 10682-784 10682-750 10682-760 10682-782 10682-780 10682-790 10682-758 10682-768 10682-778 10682-756 10682-788 10682-766 10682-754 10682-776 10682-764 10682-786 10682-752
VWR® C-Leg C-Leg Bench Frames
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Ordering information: Frame only; no worksurface included. Contact your Avantor representative for more information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty