Would you like to log in again?
Stainless Steel Wire Cage Lids, Tecniplast®
Supplier: Tecniplast
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1354G117 1354G144 1264C116 2000P962 1284L116 2154F114 1264C116 1144B116 1144B116 1284L116 2000P242 2000P214 2000P214 1354G115
77322-400EA 340.72 USD InStock
77322-400 77322-376 77322-366 77322-410 77322-426 77322-402 77322-414 10713-146 10713-102 77322-386 21020-309 10713-270 21020-308 10713-340
Stainless Steel Wire Cage Lids, Tecniplast®
Animal Supplies Animal Cages
Desgined to ensure a perfect fit and secure lock throughout the complete range of Tecniplast® cages.
Stainless steel drop-in hoppers are autoclavable. A central divider prevents the animal from crawling into the hoppper, which can be used with or without a raised wire floor.
Secure and easy to handle, these autoclavable stainless steel wire lids have cross-wire members to prevent bending. With the exception of flat lids 95057-874 and 95057-888, all lids feature a U-shaped hopper for easy access to food. Hoppers won’t entrap food or contaminants. Wire spacing is 7 mm for most sizes; 19 mm spacing for guinea pig cage lid 10713-340. Hopper divider folds down, allowing lids to be stacked for storage. Flat lids 95057-874, 95057-888, and 10713-345 are for use without a raised floor. Incorporate stopper guard for water bottle sipper tube.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty