Would you like to log in again?
Mercury Floor Machines, PRO-175, Essendant LLC FKA Lagasse
Supplier: Essendant
MFMPRO17 MFMPRO21
10714-118EA 2512.32 USD InStock
10714-118 10713-964
Mercury Floor Machines, PRO-175, Essendant LLC FKA Lagasse
Tool Sets
Precision-balanced, heavy-duty 66-frame motor ensures reliable, professional-quality performance.
  • Triple-planetary, high-torque transmission
  • Safety-enhanced interlock switch on the handle leaves both hands conveniently in control
  • Handle is fully adjustable and is comfortable for all operators
  • 50' cord length
  • 175 rpm (top speed)
  • 1.5 horsepower
  • 16" brush diameter
Triple-planetary, high-torque transmission stands up to tough tasks, while minimizing gear wear. Safety-enhanced, interlocking auto-cutoff switch enables both left- and right-hand control. Nonmarring, full-wrap bumpers protect walls and baseboards from scuffs and scrapes. Fully adjustable handle accommodates nearly any user. Smooth-rolling 6" wheels simplify transport.
Certifications: UL listed.
Ordering information: Pads and pad drivers (Sold Separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty