Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Professional Brawny Industrial® Medium Duty Premium DRC Wipers, Georgia Pacific®
Supplier: Georgia Pacific
GPC2007003CT
10714-054CS 383.59 USD InStock
10714-054
Professional Brawny Industrial® Medium Duty Premium DRC Wipers, Georgia Pacific®
Wipes Disposable Wipes and Towels Wipes for General Applications
Medium duty premium DRC wipers are strong, versatile wipers with excellent bulk, softness and absorption.
  • Cleans both water- and oil-based spills
  • Both general purpose and lab use
  • Performs like a rag without the expense
  • Tall box for easy dispensing
  • Works on dust, lint and liquid
  • Unscented
Dispenser box provides easy, one-handed, one-at-a-time dispensing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty