Would you like to log in again?
Stainless Steel Raised Wire Floors, Tecniplast®
Supplier: Tecniplast
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2150E150 1290D150 1264C150 2154F150 1264C150
77322-378EA 136.08 USD InStock
77322-378 10714-105 77322-358 77322-392 77322-428
Stainless Steel Raised Wire Floors, Tecniplast®
Animal Supplies Animal Cages
Stainless steel design with 30 mm elevation separates animals from bedding and waste.
Not required for standard operation but recommended for post-surgical animals or for those on restrictive diets. Help prevent foot pad swelling. Convenient handle design for easy insertion and removal. Autoclavable.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty