Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FBN1 Goat Polyclonal Antibody

Supplier: Abnova
PAB17824
10714-474EA 527.69 USD InStock
10714-474
Anti-FBN1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of FBN1.

Recommended Dilutions: ELISA: 1:2000

Type: Primary
Antigen: FBN1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope: internal
Host:
Isotype: Goat
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty