Would you like to log in again?
ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 7 to 67, Thermco Products
Supplier: Thermco
ACC018FFC ACC045FFC ACC064FFC ACC008CFC ACC015CFC ACC064CFC ACC030FFC ACC020FFC ACC058FFC ACC016CFC ACC062CFC ACC067CFC ACC059CFC ACC046FFC ACC048CFC ACC013CFC ACC046CFC ACC054FFC ACC029CFC ACC066CFC ACC028CFC ACC060CFC ACC020CFC ACC064CFC ACC019FFC ACC056CFC ACC016FFC ACC021CFC ACC047FFC ACC012CFC ACC047CFC ACC060FFC ACC062FFC ACC052CFC ACC063FFC ACC008FFC ACC044CFC ACC066FFC ACC054CFC ACC029FFC ACC065CFC ACC026CFC ACC007CFC ACC044FFC ACC021FFC ACC067FFC ACC007FFC ACC028FFC ACC017FFC ACC065FFC ACC050FFC ACC045CFC ACC022FFC ACC063CFC ACC059FFC ACC015FFC ACC012FFC ACC051FFC ACC017CFC ACC019CFC ACC018CFC ACC048FFC ACC056FFC ACC058CFC ACC022CFC
10747-464EA 234.89 USD InStock
10747-464 10747-420 10747-542 10747-586 10747-466 10747-588 10747-544 10747-468 10747-546 10747-590 10747-470 10747-472 10747-474 10747-574 10747-530 10747-454 10747-576 10747-532 10747-456 10747-534 10747-578 10747-458 10747-414 10747-536 10747-416 10747-538 10747-418 10143-072 10747-580 10747-460 10747-582 10747-462 10747-584 10747-540 10747-486 10747-444 10747-488 10747-840 10747-842 10747-446 10747-568 10747-448 10747-846 10747-528 10747-848 10747-490 10747-492 10747-570 10747-494 10747-450 10747-572 10747-452 10747-552 10747-554 10747-434 10747-556 10747-832 10747-436 10747-558 10747-438 10747-834 10747-838 10747-482 10747-560 10747-562
ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 7 to 67, Thermco Products
Thermometers Analog Thermometers
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil & petroleum testing.
These thermometers were designed for specific testing and can be calibrated to your specific requirements.
Certifications: A NIST Traceable Certificate of Calibration from Thermco Products ISO/IEC accredited laboratory at the specified certification points.
Ordering information: Accessories not included.
Caution: WARNING—Mercury has been designated by EPA and many state agencies as a hazardous material that can cause central nervous system, kidney and liver damage. Mercury, or its vapor, may be hazardous to health and corrosive to materials. Caution should be taken when handling mercury and mercury containing products. See the applicable product Safety Data Sheet (SDS) for details.
Order Now
ORDER
SERVICES
Services: VWRCATALYST™ offers recertification, calibration, and repair services for all makes and models of thermometers. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for quote today.
Our thermometer services include: Complete visual inspection for any physical damage. If possible, the damage will be repaired. Recertification from 1 Point to 15 Points. Thermometer readings are compared against NIST standards in high precision calibration baths, using the comparison method at determined test temperatures. Each temperature is recorded to 1/10th of a scale division. A calibration report is generated, detailing pertinent information including the correction factors for each test point. All calibration information is entered into a data storage system. All certification procedures are performed in accordance with NIST and ASTM guidelines.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty