Would you like to log in again?
ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 68 to 136, Thermco Products
Supplier: Thermco
ACC128FFC ACC068FFC ACC117CFC ACC108FFC ACC113CFC ACC123CFC ACC136FFC ACC070CFC ACC077FFC ACC097CFC ACC109FFC ACC073CBLSFC ACC130CFC ACC073FBLSFC ACC128CFC ACC069CFC ACC132CFC ACC078FFC ACC072CFC ACC114CFC ACC110CFC ACC127CFC ACC079FFC ACC126CFC ACC129CFC ACC111CFC ACC124CFC ACC098FFC ACC113FFC ACC118FFC ACC070FFC ACC080FFC ACC068CFC ACC069FFC ACC121CFC ACC112CFC ACC136CFC ACC129FFC ACC081FFC ACC110FFC ACC120CFC ACC097FFC ACC126FFC ACC130FFC ACC072FFC ACC125CFC ACC118CFC ACC116CFC ACC098CFC
10747-740EA 409.1 USD InStock
10747-740 10747-742 10747-744 10747-624 10747-702 10747-626 10747-704 10747-628 10747-706 10747-708 10747-592 10747-594 10747-696 10747-850 10747-730 10747-698 10747-852 10747-610 10747-732 10747-612 10747-734 10747-614 10747-736 10747-738 10747-684 10747-686 10747-720 10747-600 10747-688 10747-722 10747-602 10747-724 10747-726 10747-728 10747-608 10747-690 10747-692 10747-694 10747-596 10747-630 10747-598 10747-632 10747-754 10747-678 10747-756 10747-714 10747-716 10747-680 10747-682
ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 68 to 136, Thermco Products
Thermometers Analog Thermometers
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil & petroleum testing.
These thermometers were designed for specific testing and can be calibrated to your specific requirements.
Certifications: A NIST Traceable Certificate of Calibration from Thermco Products ISO/IEC accredited laboratory at the specified certification points.
Ordering information: Accessories not included.
Caution: WARNING—Mercury has been designated by EPA and many state agencies as a hazardous material that can cause central nervous system, kidney and liver damage. Mercury, or its vapor, may be hazardous to health and corrosive to materials. Caution should be taken when handling mercury and mercury containing products. See the applicable product Safety Data Sheet (SDS) for details.
Order Now
ORDER
SERVICES
Services: VWRCATALYST™ offers recertification, calibration, and repair services for all makes and models of thermometers. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for quote today.
Our thermometer services include: Complete visual inspection for any physical damage. If possible, the damage will be repaired. Recertification from 1 Point to 15 Points. Thermometer readings are compared against NIST standards in high precision calibration baths, using the comparison method at determined test temperatures. Each temperature is recorded to 1/10th of a scale division. A calibration report is generated, detailing pertinent information including the correction factors for each test point. All calibration information is entered into a data storage system. All certification procedures are performed in accordance with NIST and ASTM guidelines.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty