Would you like to log in again?
Shandon™ Cartilage Straight Thumb Forceps, Thermo Scientific
Supplier: Thermo Scientific
163
10747-996EA 19.38 USD InStock
10747-996
Shandon™ Cartilage Straight Thumb Forceps, Thermo Scientific
Forceps
Choose 12.7 cm (5") cartilage thumb forceps with straight arms and fine points for quality and precision.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship
Forceps are available in premium and standard grades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty