Would you like to log in again?
TOMO® Adhesion Microscope Slides, Matsunami Glass
Supplier: Matsunami Glass
TOMO®
TOM-13 TOM-14 TOM-15 TOM-12 TOM-11 TOM-1190 ETOM-1190
10748-174CS 602.25 USD InStock
10748-174 10748-172 10748-170 10000-038 76635-424 10748-168 10748-166
TOMO® Adhesion Microscope Slides, Matsunami Glass
Slides Adhesive Slides
Matsunami TOMO® hydrophilic adhesion slides offer superior tissue adhesion, staining performance, and slide surface fluidics
Matsunami microscope slides are manufactured in Japan using optically clear, exceptionally flat, low fluorescence glass. The slides are finished with clipped or 90° corners.
Proprietary slide coating ensures consistent performance, even after exposure to heat stress and humid environmental conditions. Recommended for use on Ventana Benchmark staining platform and automated IHC platforms.
Certifications: ISO 13485 Certified. IVD CE Marked.
Ordering information: Slides are packaged 100 per box, 10 boxes per case (1000 slides).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty