Would you like to log in again?
MAS-GP Hydrophilic Adhesion Microscope Slides, Matsunami Glass
Supplier: Matsunami Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SUMGP13 SUMGP14 SUMGP15 SUMGP11 SUMGP12
10748-176CS 445.35 USD InStock
10748-176 10748-184 10748-182 10748-180 10748-178
MAS-GP Hydrophilic Adhesion Microscope Slides, Matsunami Glass
Slides Adhesive Slides
The Matsunami MAS-GP slide surface is treated with a coating that creates a strong adhesive and hydrophilic surface
Matsunami microscope slides are manufactured in Japan using optically clear, exceptionally flat, low fluorescence glass. They are finished with clipped corners.
The hydrophilic surface ensures faster draining of the water between the glass surface and tissue section, thereby minimizing any water trapped when the tissue section is retrieved from a water bath. Reducing or eliminating the trapped water increases the adhesive bond between the tissue section and the microscope glass. The proprietary coating produces a strong positive charge on the slide. Tissue sections having a negative charge are attracted to the surface of the glass. The bond is highly durable to thermal treatment.
Ordering information: Slides are packaged 100 per box, 10 boxes per case (1000 slides).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty