Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TMEM18 Rabbit Polyclonal Antibody

Supplier: ProSci
5043_S
10750-432EA 179.85 USD InStock
10750-432
Anti-TMEM18 Rabbit Polyclonal Antibody
Antibodies
TMEM18 Antibody: Recent studies have examined how overexpression of certain genes enhances the tropism of neural stem cells (NSCs) for gliomas. Overexpression of Transmembrane protein 18 (TMEM18) provides NSCs and neural precursors an increased migration capacity towards glioblastoma cells in vitro and in the rat brain, while functional inactivation of the gene resulted in an almost complete loss of migration activity, suggesting that TMEM18 could be a potential target in NSC-based glioma therapy. Other recent results suggest that among others, the TMEM18 loci is associated with higher than normal body mass index (BMI) and obesity, indicating there may be a CNS-related role in predisposition to obesity. Multiple isoforms of TMEM18 are known to exist.

Type: Primary
Antigen: TMEM18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty