Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GATA3 Rabbit Polyclonal Antibody

Supplier: ProSci
5599_S
10750-814EA 179.85 USD InStock
10750-814
Anti-GATA3 Rabbit Polyclonal Antibody
Antibodies
GATA3 Antibody: GATA3 is a zinc finger transcription factor that was first recognized as a possible determinant of T cell development. Later studies demonstrated that it is expressed in trophoblast giant cells and regulates trophoblast-specific genes during development. Specifically, GATA3 is selectively expressed in the trophectoderm of the peri-implantation embryo and regulates CDX2, which is required for the transcriptional repression of the POU5F1/Oct4 and NANOG genes and is thus essential for the segregation of the inner cell mass and trophectoderm at the blastocyst stage. Decreased or null-expression expression of GATA3 has also been suggested to play a major role in the development and progression of luminal breast cancer.

Type: Primary
Antigen: GATA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty