Would you like to log in again?
Leica DMi1 Inverted Microscope, Leica Microsystems
Supplier: Leica
Leica®
11526208 11526208
10752-392EA 3867.44 USD InStock
CA10752-392 10752-392
Leica DMi1 Inverted Microscope, Leica Microsystems
Microscopes
Get what you need and increase efficiency in your live cell imaging workflow with the new Leica DMi1 inverted microscope.
  • Easy to use
  • Easy phase contrast
  • Cool, color-safe LED illumination
The unique flexibility of the Leica DMi1 allows you to configure the microscope for your needs with flexible condenser options; creating a solution that is just right for your lab.
Easy-to-use operation that requires minimal training and maintenance so you can fully concentrate on your research. Cool, color-safe LED illumination for constant color temperature through all stages of intensity.
Ordering information: Accessories not included. Two-week approximate lead time. Please contact your local Avantor representative if more expedited delivery times are needed.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty