Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Economy Shelf Bins, Quantum Storage Systems

Supplier: Quantum Storage Systems

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

QSB102GN QSB102IV QSB201YL QSB109BK QSB103GN QSB110GN QSB110RD QSB103BL QSB104BL QSB109IV QSB202YL QSB209BL QSB209RD QSB109YL QSB110BL QSB101GN QSB207BL QSB108IV QSB110IV QSB104BK QSB110BK QSB104IV QSB204YL QSB107GN QSB204RD QSB107IV QSB202RD QSB103YL QSB107BK QSB108RD QSB102BL QSB107BL QSB108BL QSB108GN QSB204BL QSB210YL QSB103IV QSB104YL QSB102YL QSB207YL QSB107RD QSB203RD QSB208BL QSB104RD QSB109RD QSB104GN QSB101BK QSB101RD QSB203YL QSB101YL QSB210RD QSB108YL QSB208RD QSB102BK QSB101IV QSB201BL QSB110YL QSB102RD QSB202BL QSB207RD QSB210BL QSB208YL QSB201RD QSB109BL QSB103BK QSB209YL QSB107YL QSB101BL QSB108BK QSB203BL QSB103RD QSB109GN
10752-728CS 223.77 USD InStock
10752-728 10752-528 10752-526 10752-724 10752-602 10752-612 10752-534 10752-578 10752-576 10752-532 10752-574 10752-530 10752-572 10752-570 10752-618 10752-616 10752-538 10752-736 10752-614 10752-536 10752-734 10752-546 10752-666 10752-622 10752-544 10752-664 10752-620 10752-542 10752-662 10752-580 10752-708 10752-628 10752-626 10752-548 10752-624 10752-668 10752-678 10752-634 10752-710 10752-556 10752-676 10752-632 10752-554 10752-598 10752-630 10752-674 10752-552 10752-672 10752-550 10752-670 10752-718 10752-716 10752-638 10752-714 10752-636 10752-558 10752-712 10752-568 10752-524 10752-722 10752-600 10752-644 10752-720 10752-566 10752-522 10752-642 10752-564 10752-640 10752-562 10752-682 10752-560 10752-680
Economy Shelf Bins, Quantum Storage Systems
Boxes Dispenser Boxes
Durable bins feature a mold-in label holder, built-in rear hanglock which allows bins to tilt out for complete access when on shelving. Seven divider slots.

  • Edges are reinforced for added strength
  • Designed for 12", 18" and 24" deep shelf units, racks, and standard shelving
  • Wide hopper front allows complete access to the contents of the bin
  • Available in six colors in the 4" height; Blue, Green, Ivory, Red,Yellow, and Black
  • Available in red, blue or yellow in the 6" high bin
  • Bin height available of 4" or 6"

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty