Would you like to log in again?
Stack and Nest Totes, Quantum Storage Systems
Supplier: Quantum Storage Systems
SNT180RD SNT230GY SNT225BL SNT200GY SNT185GY SNT300RD SNT180GY SNT240GY SNT200BL SNT300BL SNT195GY SNT230BL SNT190BL SNT230RD SNT300GY SNT190RD SNT185BL SNT195RD SNT225GY SNT185RD SNT180BL SNT225RD SNT200RD SNT195BL SNT190GY SNT240RD SNT240BL
10752-748CS 204.68 USD InStock
10752-748 10752-768 10752-746 10752-754 10752-698 10752-774 10752-752 10752-696 10752-772 10752-750 10752-694 10752-770 10752-692 10752-690 10752-738 10752-758 10752-756 10752-766 10752-744 10752-688 10752-742 10752-764 10752-686 10752-740 10752-762 10752-684 10752-760
Stack and Nest Totes, Quantum Storage Systems
Boxes Storage Boxes
Heavy-duty and dependable stack and nest totes are reusable container for storing and transporting your valuable items in any application.
  • Available in red, blue, or gray
  • Available in nine sizes
  • Unique ability to stack or they can be turned 180o to nest when empty saving valuable space without using a lid
  • Lids available
Injection molded from high-density polypropylene that complies with the FDA and USDA regulations.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty