Would you like to log in again?
Halogen Moisture Analyzer HE73, METTLER TOLEDO®
Supplier: Mettler-Toledo
30237205 30237205
10753-188EA 4710 USD InStock
10753-188 CA10753-142
Halogen Moisture Analyzer HE73, METTLER TOLEDO®
Analyzers Moisture Analyzers
The HE73 Halogen Moisture Analyzer is a reliable and cost-effective solution for determining moisture content.
  • Reliable and cost-effective
  • Simple to use
  • Three-step process
  • Metal base
  • Stainless steel surface
The instrument is simple to use, thanks to straightforward navigation and on screen guidance.
Additionally, the HE73 includes two programmable methods and added flexibility thanks to additional switch-off criterion and a higher capacity.
Proper installation and initial calibration are the key to ensuring the moisture analyzer performs properly from day one. A METTLER TOLEDO® authorized service technician performs installation qualification and operational qualification (IQ/OQ).
Simply follow the easy, three-step process that the instrument walks users through and achieve reliable moisture readings in minutes. A metal base and easy-to-clean stainless steel surface ensure this a product that is built to last.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: METTLER TOLEDO® provides several service options, including Installation and Qualification, as well as extended warrantys. To learn more about these services please search "METTLER TOLEDO® Installation and Qualification Services" in the website search field.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty