Would you like to log in again?
Thermal Mixer II, Boekel Scientific
Supplier: Boekel Scientific
270900 270800
10753-396EA 5222.07 USD InStock
10753-396 10753-398
Thermal Mixer II, Boekel Scientific
Shakers and Mixers Shaking Incubators
The Boekel Scientific line of Thermal Mixers consists of a Heat/Cool unit and a Heat only unit
  • Large color touch screen
  • Programmable heating, cooling and mixing sequences
  • Auto recognized blocks with 11 Interchangeable
  • Program up to five independent steps
  • Simple, accurate four point calibration
  • Easy to remove blocks
Easy to use and program due to its large, intuitive color touch screen. Both the Heat/Cool and Heat Only units contain an auto-recognition feature that identifies the block placed on the unit and Boekel offers a variety of blocks to accommodate numerous tube sizes, microplates and PCR plates.
The user can easily view and access all parameters from the Home Screen and instantly change those parameters for a step, if need be. Each unit stores up to 25 - 5 step programs and the user can easily name each program for easy process recall.
Each unit is designed for precise sample preparation when working with DNA, RNA, plasmids, cultures and anzymatic reactions.
Certifications: Unit is powered via an AC Adapter that is UL, CSA and CE certified.
Ordering information: Each unit is supplied with a block for 24×1.5 ml tubes and carries a two-year manufacturers warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty