Would you like to log in again?
Mycoplasma Agar with Cefoperazone, Hardy Diagnostics
Supplier: Hardy Diagnostics
G212 G212
10753-454PK 80.39 USD InStock
10753-454 CA10753-454
Mycoplasma Agar with Cefoperazone, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Mycoplasma agar with cefoperazone is used for the detection of Mycoplasma spp
associated with bovine mastitis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty