Would you like to log in again?
Advanced Touch Screen Dry Baths/Block Heaters, Thermo Scientific
Supplier: Thermo Scientific
88870008 88870008 88870009 88870009 88870007 88870007
10753-544EA 1261.77 USD InStock
10753-544 10753-594 CA10753-594 10753-546 CA10753-544 CA10753-546
Advanced Touch Screen Dry Baths/Block Heaters, Thermo Scientific
Block Heaters Digital Block Heaters
Advanced Touch Screen Dry Baths/Block Heaters are designed to achieve precise temperature stability and uniformity with an easy-to-use touch screen
  • Easy-to-use touch screen control displays time and temperature
  • Built-in temperature sensing probe designed for outstanding temperature accuracy and control
  • Temperature range: Ambient +5°C to 130°C
  • Temperature deviation adjustment
  • Precise temperature control with PID circuit
  • 10 user setting programs; each supporting up to 5 sequential steps of different temperatures and duration times
  • Temperature calibration to offset the temperature to desired value
  • Timer enables the user to accurately monitor the heating time
  • Interchangeable aluminum alloy sample blocks provide versatility and allow for easy cleaning and disinfecting
  • Built in over-temperature protection device is designed to provide reliable operation
  • Powder-coated steel body construction ensures durability
  • Optional External Temperature Probe: when connected, the internal temperature sensor will disconnect automatically, allowing the temperature control and display to rely on the external temperature sensor
  • Two-year warranty
  • Blocks sold separately
Units offer a range of configurations to hold 1, 2, and 4 interchangeable modular blocks, accommodating a variety of vessels and applications.
Ordering information: For a comprehensive list of optional accessory items, please see Accessories for Advanced Touch Screen and Digital Dry Baths/Block Heaters, Thermo Scientific.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty