Would you like to log in again?
Analytical and Precision Balances, MS-TS
Supplier: Mettler-Toledo
MS-TS
30133545 30133544 30133528 30133523 30133551 30133534 30133541 30133532 30133529 30133525 30133528 30133531 30133547 30133522 30133549 30133525 30133531 30133524 30133550 30133532 30133526 30133527 30133552 30133535 30133533 30133543 30133530 30133535 30133529 30133548 30133523 30133533 30133527 30133536 30133546 30133536 30133526 30133524 30133522 30133530 30133534 30133542
10753-710EA 7590 USD InStock
CA10753-562 10753-710 CA10753-564 10753-698 CA10753-560 10753-570 76307-416 76307-414 76307-412 10753-708 76307-410 10753-704 10753-706 CA10753-710 10753-568 10753-700 CA10753-698 CA10753-558 CA10753-712 10753-702 76307-418 10753-564 10753-566 CA10753-570 10753-560 10753-562 76307-426 76307-424 76307-422 CA10753-706 76307-420 CA10753-708 CA10753-702 CA10753-704 10753-558 10753-712 76307-408 CA10753-566 76307-406 76307-428 CA10753-568 CA10753-700
Analytical and Precision Balances, MS-TS
Balances and Scales
METTLER TOLEDO® MS-TS series balances are reliable and robust balances designed for everyday weighing.
  • Reliable and robust
  • Easy to operate
  • Advanced security features
  • Metal housing
  • Easy to clean
Easy to operate units incorporate advanced security features, and these balances make even sophisticated weighing simple and worry free.
To ensure trusted results, the MS-TS series balances feature built-in checks and safety features. All units include MinWeigh warnings, LevelControl, ISO-Log (adjustment history), sample IDs, passcode protection, and Automatic Internal Adjustment (FACT). These easy to use units are supplied standard with a 17.8 cm (7") extra-large color TFT touchscreen, an intuitive user interface, and more than ten built-in applications. The robust and durable design features full metal housing, a robust Monobloc weighing cell, and massive overload protection. Units are easy to clean and chemical-resistant, as the full metal housing is resistant to most chemicals including acetone. A QuickLock feature for draft shields and glass panels allows dismantling in seconds.
Capacities range from 120 to 12200 g, with readabilities starting from 0.1 mg.
Certifications: UL listed and CSA certified.
Ordering information: The two-place readability models do not include draft shields. QuickLock draft shields are only supplied with models featuring three-place readability or better. Optional Service packages are offered to assure continual reliability and maintain optimum balance performance. Proper installation and initial calibration are key to ensuring the balance performs properly from day one. A METTLER TOLEDO® technician performs installation qualification and operational qualification (IQ/OQ). Contact your representative for Service package details.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: METTLER TOLEDO® provides several service options, including installation and qualification, as well as extended warrantys. To learn more about these services please search "METTLER TOLEDO® Installation and Qualification Services" in the website search field.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty