Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MS4A1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2H7]

Supplier: Tonbo Biosciences
20-0209-T100 20-0209-T025 20-0209-T500
10753-720EA 89.8 USD InStock
10753-720 10753-722 10753-724
Anti-MS4A1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2H7]
Antibodies
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 2H7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty