Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SK3]

Supplier: Tonbo Biosciences
35-0047-T100 35-0047-T025 35-0047-T500
10753-752EA 216 USD InStock
10753-752 10753-750 10753-748
Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SK3]
Antibodies
The SK3 antibody reacts with human CD4, a 59 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4, and co-receptors CCR5 and CXCR4, may also be utilized by HIV-1 to enter T cells. Human CD4 is typically expressed on thymocytes, some mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells. The SK3 antibody is widely used as a phenotypic marker for human CD4 expression, and has been reported to be cross-reactive with Rhesus and Cynomolgus CD4. This antibody does not block binding of alternative clone RPA-T4, suggesting that they recognize different epitopes.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: SK3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty