Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD5 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: L17F12]

Supplier: Tonbo Biosciences
67-0058-T025 67-0058-T100 67-0058-T500
10753-820EA 145 USD InStock
10753-820 10753-822 10753-824
Anti-CD5 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: L17F12]
Antibodies
The L17F12 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is a member of the scavenger receptor superfamily and is present on approximately 70% of normal peripheral blood lymphocytes. CD5 is involved in modulating antigen receptor signaling in both T and B cells and acts through binding of CD72, its receptor expressed on B cells.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty