Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Unisex 100% Cotton Lab Coat

Supplier: VWR
VWR®
10753-938 10753-936 10753-940 10753-932 10753-904 10753-934 10753-930
10753-930CS 960.19 USD InStock
10753-930 10753-940 10753-936 10753-938 10753-904 10753-932 10753-934
VWR® Unisex 100% Cotton Lab Coat
Workwear
These unisex cotton lab coats are made of breathable, comfortable, 100% cotton fabric and feature a quick-release five-snap front and snap cuffs

  • Quick-release five-snap front and snap cuffs
  • Plain front with no embroidery
  • Overall length is 99 cm (39")

In addition, the 100% cotton lab coat features one breast pocket with a pencil slot, plus two oversized hip pockets. Side vents are also included for easy pants pocket access.




Ordering information: Sizes XX-Small, 4X-Large, and 5X-Large are also available via special order. Contact your Avantor sales representative for more information. Please allow up to 6 weeks for delivery of special order items.

Order Now
 

ORDER

SPECIFICATIONS

COMPLEMENTARY PRODUCTS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty