Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HardyDisk™ AST , Hardy Diagnostics
Supplier: Hardy Diagnostics
Z8321 Z8575 Z8791 Z8315 Z8851 Z8325 Z8815 Z9011 Z9121 Z9245 Z8905 Z9241 Z8365 Z9121 Z8275 Z8311 Z9015 Z8241 Z9011 Z8361 Z8795 Z9241 Z9125 Z8735 Z8245 Z8411 Z8361 Z8855 Z8851
10753-972EA 33.59 USD InStock
10753-972 10753-974 10753-970 75832-516 75832-512 75832-732 75832-484 75832-486 10753-968 CA10753-974 CA10753-976 CA10753-970 CA10753-972 75832-728 75832-508 75832-504 75832-724 75832-726 75832-506 75832-488 75832-722 75832-502 75832-496 75832-498 75832-492 75832-494 75832-490 CA10753-968 10753-976
HardyDisk™ AST , Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
HardyDisk™ AST’s are impregnated paper disks used for antimicrobial susceptibility testing (AST), also known as disk diffusion or Kirby-Bauer testing
Fully compatible with the BBL single or multiple disk dispensers.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty