Would you like to log in again?
Phenylalanine Agar Slant, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
L21 L21
10753-978PK 111.23 USD InStock
CA101380-780 10753-978
Phenylalanine Agar Slant, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Phenylalanine agar slant is used for the differentiation of gram negative bacilli based on the production of phenylpyruvic acid by oxidative deamination
Use with ferric chloride 10% (cat no. Z63).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty