Would you like to log in again?
CompactDry™ ETB, Enterobacteriaceae, Hardy Diagnostics
Supplier: Hardy Diagnostics
CompactDry
ETB100 ETB100
10753-986PK 168.52 USD InStock
10753-986 CA10753-986
CompactDry™ ETB, Enterobacteriaceae, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
CompactDry™ is a ready-to-use system that reduces the time needed to perform microbial testing on food, cannabis, meat, cosmetics, raw materials and more.
  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label
The user friendly nature of the CompactDry™ plate allows for quick adoption in cannabis and food testing labs, regardless of technician experience.
Colonies from the Enterobacteriaceae group will turn red or purple due to the presence o+T14+K14:U14+J14:U14.
Ordering information: This product is not intended for use in the diagnosis of human disease.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty