Would you like to log in again?
25-Place Low Temperature Microscope Slide Box
Supplier: Heathrow Scientific
HS120316
10754-104CS 1337.82 USD InStock
10754-104
25-Place Low Temperature Microscope Slide Box
Microscope Slide Boxes
Store slide samples in ultra-low temperatures with confidence.
  • Made of durable polycarbonate to store in ultra-low temperatures
  • Uniform grooved slide slots separate slides preventing slide contact
  • Molded feet enable secure stacking for easy transport and storage
  • Low temperature resistant numbered foam lining corresponds with identification sheet
Protect slides in temperature ranges as low as –80 °C.
Lock type: Friction fit
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty