Would you like to log in again?
Corrugated Sample Storage Tray
Supplier: Heathrow Scientific
HS120375
10754-106PK 188.15 USD InStock
10754-106
Corrugated Sample Storage Tray
Trays/Dishes/Pots Laboratory Trays/Dishes
Corrugated storage trays can be used with the included dividers for specimen containers or remove them for general lab product storage or transport. Affordable sample organization and storage.
  • Made from corrugated polypropylene
  • Moisture resistant, significantly reducing the chance of mold
  • Lightweight and sturdy to hold up to 24 full specimen containers
  • Stackable with other trays
  • Removable dividers allow outer shell to be used for storage or transport of laboratory products
Ordering information: Supplied flat-packed, each order includes 10 trays and 10 dividers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty